Learn More
American Research Products Inc Pig Nuclear factor kappa B NF
Catalog No. 5000328333
Encompass
Pig Nuclear factor-kappa B NF- B ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28333 Supplier American Research Products Inc Supplier No. CSBE13448P
Pig Nuclear factor-kappa B NF- B ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich