Learn More
Biomatik Corporation Pig Osteopontin (OPN) ELISA Kit
Supplier:  Biomatik Corporation EKU06432
Encompass_Preferred
Pig Osteopontin (OPN) ELISA Kit for detecting OPN in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P14287. Gene ID: 397087. Target Synonyms: SPP1; BNSP; BSPI; ETA1; Secreted Phosphoprotein 1; Bone Sialoprotein I; Early T-Lymphocyte Activation 1; Nephropontin; Urinary stone protein; Uropontin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8826