Learn More
American Research Products Inc Pig oXidized low density lipop
Catalog No. 5000326542
Encompass
Pig oXidized low density lipoprotein OXLDL ELISA Kit from CUSABIO Detection Range 0 39 nmol/mL-100 nmol/mL Sensitivity 0 39 nmol/mL Sample serum plasma Method Competitive
Catalog No. 50-003-26542 Supplier American Research Products Inc Supplier No. CSBE06527P
Pig oXidized low density lipoprotein OXLDL ELISA Kit from CUSABIO Detection Range 0 39 nmol/mL-100 nmol/mL Sensitivity 0 39 nmol/mL Sample serum plasma Method Competitive