Learn More
Biomatik Corporation Pig Phospholipase A2, Pancreas (pPLA2) ELISA Kit
Supplier:  Biomatik Corporation EKU06625
Encompass_Preferred
Pig Phospholipase A2, Pancreas (pPLA2) ELISA Kit for detecting pPLA2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 31.25-2000pg/ml. Sensitivity: 13.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00592. Gene ID: 445525. Target Synonyms: PLA2G1B; PLA2A; PPLA2; Phospholipase A2, Group IB; Group IB phospholipase A2; Phosphatidylcholine 2-acylhydrolase 1B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5585