Learn More
American Research Products Inc Pig plasminogen actiVator inhi
Catalog No. 5000328755
Encompass
Pig plasminogen actiVator inhibitor 1 PAI-1 ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28755 Supplier American Research Products Inc Supplier No. CSBE06534P
Pig plasminogen actiVator inhibitor 1 PAI-1 ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich