Learn More
American Research Products Inc Pig Platelet DeriVed Growth Fa
Catalog No. 5000328904
Encompass
Pig Platelet-DeriVed Growth Factor AB (PDGF-AB) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28904 Supplier American Research Products Inc Supplier No. CSBEQ027978PI
Pig Platelet-DeriVed Growth Factor AB (PDGF-AB) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich