Learn More
Biomatik Corporation Pig Platelet Derived Growth Factor Subunit B (PDGFB) CLIA Kit
Supplier:  Biomatik Corporation EKU09544
Encompass_Preferred
Pig Platelet Derived Growth Factor Subunit B (PDGFB) CLIA Kit for detecting PDGFB in plasma. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.08pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: A0A287ADF0. Target Synonyms: PDGF-B; PDGF2; SIS; SSV; C-sis; Simian Sarcoma Viral (v-sis) Oncogene Homolog; Platelet Derived Growth Factor Beta Polypeptide; Proto-oncogene c-Sis; Becaplermin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8500