Learn More
Biomatik Corporation Pig Prealbumin (PALB) ELISA Kit
Supplier:  Biomatik Corporation EKU07866
Encompass_Preferred
Pig Prealbumin (PALB) ELISA Kit for detecting PALB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.56-100ng/ml. Sensitivity: 0.71ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50390. Gene ID: 397419. Target Synonyms: TBPA; PA; PLB; ATTR; TTR; Transthyretin; Prealbumin Amyloidosis Type I; Transports Thyroxine and Retinol. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6887