Learn More
Innovative Research Inc Pig RBCs Packed 10 50ml
Catalog No. 502744119
Encompass_Preferred
Porcine Red Blood Cells Packed 10% 50ml - Porcine Red Blood Cells from Innovative Research are manufactured using whole blood collections from SPECIES. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 10% suspension in Alsever's Solution. Red blood cells are perishable and are collected and processed upon receipt of order.
Catalog No. 50-274-4119 Supplier Innovative Research Inc Supplier No. IPCRBC10P50ML
Porcine Red Blood Cells Packed 10% 50ml - Porcine Red Blood Cells from Innovative Research are manufactured using whole blood collections from SPECIES. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 10% suspension in Alsever's Solution. Red blood cells are perishable and are collected and processed upon receipt of order.