Learn More
Biomatik Corporation Pig Retinol Binding Protein 4 (RBP4) ELISA Kit
Supplier:  Biomatik Corporation EKU11401
Encompass_Preferred
Pig Retinol Binding Protein 4 (RBP4) ELISA Kit for detecting RBP4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 3.12-200ng/ml. Sensitivity: 1.45ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27485. Gene ID: 397124. Target Synonyms: PRBP; RBP; Plasma retinol-binding protein; Retinol Binding Protein 4, Plasma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5394