Learn More
American Research Products Inc Pig ScaVenger receptor cystein
Catalog No. 5000327607
Encompass
Pig ScaVenger receptor cysteine-rich type 1 protein M130 (CD163) ELISA kit from CUSABIO Detection Range 93 75 ng/mL-6000 ng/mL Sensitivity 23 4 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27607 Supplier American Research Products Inc Supplier No. CSBEL004882PI
Pig ScaVenger receptor cysteine-rich type 1 protein M130 (CD163) ELISA kit from CUSABIO Detection Range 93 75 ng/mL-6000 ng/mL Sensitivity 23 4 ng/mL Sample serum plasma tissue homogenates Method Sandwich