Learn More
American Research Products Inc Pig secretory immunoglobulin A
Catalog No. 5000329198
Encompass
Pig secretory immunoglobulin A SIgA ELISA Kit from CUSABIO Detection Range 0 15 g/mL-37 5 g/mL Sensitivity 0 15 g/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-29198 Supplier American Research Products Inc Supplier No. CSBE12063P
Pig secretory immunoglobulin A SIgA ELISA Kit from CUSABIO Detection Range 0 15 g/mL-37 5 g/mL Sensitivity 0 15 g/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive