Learn More
Biomatik Corporation Pig Somatostatin (SST) ELISA Kit
Supplier:  Biomatik Corporation EKU07412
Encompass_Preferred
Pig Somatostatin (SST) ELISA Kit for detecting SST in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 6.17-500pg/ml. Sensitivity: 2.74pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01168. Gene ID: 494469. Target Synonyms: GHIH; SRIF; SMST; GHRIH; GHRIF; Growth Hormone Releasing Inhibiting Hormone; Somatotropin Release-Inhibiting Factor; Somatostatin-14; Somatostatin-28. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8752