Learn More
American Research Products Inc Pig Stromal cell deriVed facto
Catalog No. 5000329242
Encompass
Pig Stromal cell deriVed factor 1a SDF-1a ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma cell culture supernates tissue homogenates cell Lysates Method Sandwich
Catalog No. 50-003-29242 Supplier American Research Products Inc Supplier No. CSBE12055P
Pig Stromal cell deriVed factor 1a SDF-1a ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma cell culture supernates tissue homogenates cell Lysates Method Sandwich