Learn More
Biomatik Corporation Pig Superoxide Dismutase 1 (SOD1) ELISA Kit
Supplier:  Biomatik Corporation EKU09097
Encompass_Preferred
Pig Superoxide Dismutase 1 (SOD1) ELISA Kit for detecting SOD1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04178. Target Synonyms: ALS; ALS1; IPOA; Superoxide Dismutase 1, Soluble; Amyotrophic Lateral Sclerosis 1,Adult; Superoxide dismutase [Cu-Zn]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4473