Learn More
American Research Products Inc Pig terminal complement comple
Catalog No. 5000330139
Encompass
Pig terminal complement compleX C5b-9 (TCC C5b-9) ELISA Kit from CUSABIO Detection Range 18 75 ng/mL-1200 ng/mL Sensitivity 4 69 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30139 Supplier American Research Products Inc Supplier No. CSBE15837P
Pig terminal complement compleX C5b-9 (TCC C5b-9) ELISA Kit from CUSABIO Detection Range 18 75 ng/mL-1200 ng/mL Sensitivity 4 69 ng/mL Sample serum plasma tissue homogenates Method Sandwich