Learn More
Biomatik Corporation Pig Thrombospondin 1 (THBS1) ELISA Kit
Supplier:  Biomatik Corporation EKU11469
Encompass_Preferred
Pig Thrombospondin 1 (THBS1) ELISA Kit for detecting THBS1 in plasma. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 78-5,000pg/ml. Sensitivity: 32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1SS26. Gene ID: 492313. Target Synonyms: TSP1; Thrombospondin-1p180. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6018