Learn More
American Research Products Inc Pig Thyroid Stimulating Hormon
Catalog No. 5000329398
Encompass
Pig Thyroid Stimulating Hormone TSH ELISA Kit from CUSABIO Detection Range 0 6 IU/mL-24 IU/mL Sensitivity 0 3 IU/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29398 Supplier American Research Products Inc Supplier No. CSBE13719P
Pig Thyroid Stimulating Hormone TSH ELISA Kit from CUSABIO Detection Range 0 6 IU/mL-24 IU/mL Sensitivity 0 3 IU/mL Sample serum plasma Method Sandwich