Learn More
Biomatik Corporation Pig Thyrotropin Releasing Hormone (TRH) ELISA Kit
Supplier:  Biomatik Corporation EKU11577
Encompass_Preferred
Pig Thyrotropin Releasing Hormone (TRH) ELISA Kit for detecting TRH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.93pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P62968. Target Synonyms: TRF; Thyrotropin-Releasing Factor; Thyroliberin; Protirelin; Pro-tirelin; Prothyroliberin; TSH-releasing factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7000