Learn More
Biomatik Corporation Pig Tissue Factor (TF) ELISA Kit
Supplier:  Biomatik Corporation EKU07698
Encompass_Preferred
Pig Tissue Factor (TF) ELISA Kit for detecting TF in plasma. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6RCS8. Gene ID: 396677. Target Synonyms: CD142; FIII; F3; TFA; Thromboplastin; Coagulation Factor III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6661