Learn More
American Research Products Inc Pig tissue inhibitors of metal
Catalog No. 5000328962
Encompass
Pig tissue inhibitors of metalloproteinase 1 TIMP-1 ELISA Kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28962 Supplier American Research Products Inc Supplier No. CSBE06799P
Pig tissue inhibitors of metalloproteinase 1 TIMP-1 ELISA Kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum plasma tissue homogenates Method Sandwich