Learn More
Biomatik Corporation Pig Tissue Inhibitors Of Metalloproteinase 4 (TIMP4) CLIA Kit
Supplier:  Biomatik Corporation EKU09904
Encompass_Preferred
Pig Tissue Inhibitors Of Metalloproteinase 4 (TIMP4) CLIA Kit for detecting TIMP4 in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 13.7-10,000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9TTB6. Target Synonyms: TIMP Metallopeptidase Inhibitor 4; Tissue Inhibitor of Metalloproteinases 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5974