Learn More
American Research Products Inc Pig transforming growth factor
Catalog No. 5000330194
Encompass
Pig transforming growth factor b1 (TGF-b1) ELISA Kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-30194 Supplier American Research Products Inc Supplier No. CSBE06843P
Pig transforming growth factor b1 (TGF-b1) ELISA Kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma Method Sandwich