Learn More
Biomatik Corporation Pig Trypsin (TRY) CLIA Kit
Supplier:  Biomatik Corporation EKU09454
Encompass_Preferred
Pig Trypsin (TRY) CLIA Kit for detecting TRY in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.27-200ng/ml. Sensitivity: 0.11ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P00761. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6245