Learn More
Cloud Clone Corp Pig Trypsin (TRY) (Sandwich ELISA) ELISA Kit
Catalog No. NC4011238
Encompass_Preferred
Pig Trypsin (TRY) (Sandwich ELISA) ELISA Kit measures Pig TRY in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids.This assay has high sensitivity and excellent specificity for detection of Trypsin (TRY).No significant cross-reactivity or interference between Trypsin (TRY) and analogues was observed.Detection Range: 1.56-100ng/mLSensitivity:The minimum detectable dose of this kit is typically less than 0.57ng/mLDetection menthod: ColormetricGene ID:-
Catalog No. NC4011238 Supplier Cloud Clone Corp Supplier No. SEA250PO
Pig Trypsin (TRY) (Sandwich ELISA) ELISA Kit measures Pig TRY in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids.This assay has high sensitivity and excellent specificity for detection of Trypsin (TRY).No significant cross-reactivity or interference between Trypsin (TRY) and analogues was observed.Detection Range: 1.56-100ng/mLSensitivity:The minimum detectable dose of this kit is typically less than 0.57ng/mLDetection menthod: ColormetricGene ID:-