Learn More
American Research Products Inc Pig Vascular Endothelial cell
Catalog No. 5000329767
Encompass
Pig Vascular Endothelial cell Growth Factor VEGF ELISA KIT from CUSABIO Detection Range 4 69 pg/mL-300 pg/mL Sensitivity 1 17 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29767 Supplier American Research Products Inc Supplier No. CSBE12053P
Pig Vascular Endothelial cell Growth Factor VEGF ELISA KIT from CUSABIO Detection Range 4 69 pg/mL-300 pg/mL Sensitivity 1 17 pg/mL Sample serum plasma tissue homogenates Method Sandwich