Learn More
Biomatik Corporation Pig Vascular Endothelial Growth Factor Receptor 1 (VEGFR1) ELISA Kit
Supplier:  Biomatik Corporation EKU08104
Encompass_Preferred
Pig Vascular Endothelial Growth Factor Receptor 1 (VEGFR1) ELISA Kit for detecting VEGFR1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: ACH73486.1. Target Synonyms: Flt1; Fms-Related Tyrosine Kinase 1; Vascular Permeability Factor Receptor; Fms-like tyrosine kinase 1; Tyrosine-protein kinase receptor FLT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4554