Learn More
Aviva Systems Biology Corp PIGF Antibody - N-terminal region (ARP40408_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40408_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIGF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PIGF. Alternate names: MGC32646, MGC33136ApplicationWB, IHC. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat, Sheep
Catalog No. 50-014-57435