Learn More
Aviva Systems Biology Corp PIGK Antibody - N-terminal region (ARP40588_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40588_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIGK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PIGK. Alternate names: GPI8, MGC22559ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57536