Learn More
Aviva Systems Biology Corp PIGL Antibody - N-terminal region (ARP46427_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46427_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIGL. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PIGL. Alternate names: CHIMEApplicationWB. Species reactivity: Human
Catalog No. 50-285-257