Learn More
Creative Biomart Pigment Epithelium-Derived Factor (PEDF)
Catalog No. 502084324
Encompass_Preferred
PEDF Human Recombinant produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 400 amino acids and having a molecular mass of 44.5 kDa. The Human PEDF is purified by proprietary chromatographic techniques.
Catalog No. 50-208-4324 Supplier Creative Biomart Supplier No. THP0273
PEDF Human Recombinant produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 400 amino acids and having a molecular mass of 44.5 kDa. The Human PEDF is purified by proprietary chromatographic techniques.