Learn More
BURKLE INC Pigment lance, flute open. 50%, flute 35.43 in. (900mm), 9.23 oz. (273ml)
Catalog No. 5000025871
Encompass_Preferred
Pigment lance to sample pigments, pastes and agglomerating bulk goods.
Catalog No. 50-000-25871 Supplier BURKLE INC Supplier No. 53690910
Pigment lance to sample pigments, pastes and agglomerating bulk goods.