Learn More
BURKLE INC Pigment lance, flute open. 50%, flute 11.81 in. (300 mm), 2.87 oz. (85 ml)
Catalog No. 5000026052
Encompass_Preferred
Pigment lance to sample pigments, pastes and agglomerating bulk goods.
Catalog No. 50-000-26052 Supplier BURKLE INC Supplier No. 53690310
Pigment lance to sample pigments, pastes and agglomerating bulk goods.