Learn More
BURKLE INC Pigment lance, flute open.25%, length 11.81 in. (300 mm), 4.90 oz. (145 ml)
Catalog No. 5000025701
Encompass_Preferred
Pigment lance to sample pigments, pastes and agglomerating bulk goods.
Catalog No. 50-000-25701 Supplier BURKLE INC Supplier No. 53690300
Pigment lance to sample pigments, pastes and agglomerating bulk goods.