Learn More
BURKLE INC Pigment lance, flute open. 50%, flute 23.62 in. (600mm), 6.05 oz. (179ml)
Catalog No. 5000025702
Encompass_Preferred
Pigment lance to sample pigments, pastes and agglomerating bulk goods.
Catalog No. 50-000-25702 Supplier BURKLE INC Supplier No. 53690610
Pigment lance to sample pigments, pastes and agglomerating bulk goods.