Learn More
BURKLE INC Pigment lance, flute open. 25%, flute 23.62 in. (600mm), 10.54 oz. (312ml)
Catalog No. 5000025869
Encompass_Preferred
Pigment lance to sample pigments, pastes and agglomerating bulk goods.
Catalog No. 50-000-25869 Supplier BURKLE INC Supplier No. 53690600
Pigment lance to sample pigments, pastes and agglomerating bulk goods.