Learn More
Medchemexpress LLC Pigment Yellow 101 | 2387-03-3 | 98.06% | 50 MG
SDP
Catalog No. 5000410432
Encompass_Preferred
Pigment Yellow 101 is a yellow bis-azomethine pigment that exhibits solid-state fluorescence with excitation/emission at 411 nm/512 nm. This product is available as a commercial colorant.
  • Yellow bis-azomethine pigment
  • Excitation/emission: 411 nm/512 nm
  • Exhibits solid-state fluorescence
  • Available as a commercial colorant
Catalog No. 50-004-10432 Supplier Medchemexpress LLC Supplier No. HYD032450MG
Pigment Yellow 101 is a yellow bis-azomethine pigment that exhibits solid-state fluorescence with excitation/emission at 411 nm/512 nm. This product is available as a commercial colorant.
  • Yellow bis-azomethine pigment
  • Excitation/emission: 411 nm/512 nm
  • Exhibits solid-state fluorescence
  • Available as a commercial colorant