Learn More
Medchemexpress LLC Pigment Yellow 101 | 2387-03-3 | 98.1% | 100 MG
SDP
Catalog No. 5000410403
Encompass_Preferred
Pigment Yellow 101 is a yellow bis-azomethine pigment that exhibits solid-state fluorescence and is available as a commercial colorant.
  • Exhibits solid-state fluorescence
  • Available as a commercial colorant
  • Yellow bis-azomethine pigment
  • Excitation/emission: 411 nm/512 nm
Catalog No. 50-004-10403 Supplier Medchemexpress LLC Supplier No. HYD0324100MG
Pigment Yellow 101 is a yellow bis-azomethine pigment that exhibits solid-state fluorescence and is available as a commercial colorant.
  • Exhibits solid-state fluorescence
  • Available as a commercial colorant
  • Yellow bis-azomethine pigment
  • Excitation/emission: 411 nm/512 nm