Learn More
Medchemexpress LLC Pigment Yellow 101 | 2387-03-3 | 98.1% | 25 MG
SDP
Catalog No. 5000413592
Encompass_Preferred
Pigment Yellow 101 is a yellow bis-azomethine pigment (excitation/emission: 411 nm/512 nm) that exhibits solid-state fluorescence and is available as a commercial colorant. It is suitable for laboratory centrifugation and storage.
  • Excitation/emission: 411 nm/512 nm
  • Exhibits solid-state fluorescence
  • For research use only
Catalog No. 50-004-13592 Supplier Medchemexpress LLC Supplier No. 25MGHYD0324
Pigment Yellow 101 is a yellow bis-azomethine pigment (excitation/emission: 411 nm/512 nm) that exhibits solid-state fluorescence and is available as a commercial colorant. It is suitable for laboratory centrifugation and storage.
  • Excitation/emission: 411 nm/512 nm
  • Exhibits solid-state fluorescence
  • For research use only