Learn More
Aviva Systems Biology Corp PIGO Antibody - N-terminal region (ARP50093_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50093_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIGO. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PIGO. Alternate names: DKFZp434M222, FLJ00135, MGC20536, MGC3079, RP11-182N22.4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60259