Learn More
MSC PIGSKIN HD MIG GLOVES
Catalog No. 5000787652
Encompass
This top grain pigskin unlined MIG glove is naturally resistant to moisture, oils, and water. It features a straight thumb for an enhanced feel and gun grip, and a 4-inch cuff for added protection. The glove is sewn with Kevlar for increased strength.
  • Top grain pigskin is naturally resistant to oils and water
  • Straight thumb for a great feel and gun grip
  • Provides 4-inch cuff for added protection
  • Sewn with Kevlar for added strength
  • Available in vend-ready packaging
Catalog No. 50-007-87652 Supplier MSC Supplier No. 00719575
This top grain pigskin unlined MIG glove is naturally resistant to moisture, oils, and water. It features a straight thumb for an enhanced feel and gun grip, and a 4-inch cuff for added protection. The glove is sewn with Kevlar for increased strength.
  • Top grain pigskin is naturally resistant to oils and water
  • Straight thumb for a great feel and gun grip
  • Provides 4-inch cuff for added protection
  • Sewn with Kevlar for added strength
  • Available in vend-ready packaging