Learn More
Aviva Systems Biology Corp PIGT Antibody - N-terminal region (ARP47203_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47203_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIGT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PIGT. Alternate names: CGI-06, FLJ41596, MGC8909, NDAPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59484