Learn More
Aviva Systems Biology Corp Pigu Antibody - N-terminal region (ARP50246_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50246_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pigu. It was validated on Western Blot. . Alternate names: 5430426F17Rik, BC028278, Cdc91l1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-500