Learn More
Aviva Systems Biology Corp PIGW Antibody - middle region (ARP49147_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49147_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIGW. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PIGW. Alternate names: FLJ37433, Gwt1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-60031