Learn More
Aviva Systems Biology Corp Pigw Antibody - middle region (ARP49146_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49146_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pigw. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: PigwApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-285-401