Learn More
Aviva Systems Biology Corp PIGX Antibody - C-terminal region (ARP60842_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60842_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIGX. It was validated on Western Blot. . Alternate names: FLJ20522ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-287-187