Learn More
Aviva Systems Biology Corp PIGZ Antibody - N-terminal region (ARP41020_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41020_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PIGZ. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PIGZ. Alternate names: FLJ12768, GPI-MT-IV, MGC52163, SMP3, PIG-ZApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-57698