Learn More
Medchemexpress LLC HY-10303 5mg Medchemexpress, PIK-294 CAS:900185-02-6 Purity:>98%
SDP
Catalog No. 501968023
Encompass_Preferred
Medchemexpress, HY-10303 5mg PIK-294 CAS:900185-02-6 PIK-294 is a potent p110δ-selective inhibitor with an IC50 of 10 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8023 Supplier Medchemexpress LLC Supplier No. HY103035MG
Medchemexpress, HY-10303 5mg PIK-294 CAS:900185-02-6 PIK-294 is a potent p110δ-selective inhibitor with an IC50 of 10 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.