Learn More
Selleck Chemical LLC PIK-75 HCl 10mg 372196-77-5
SDP
Catalog No. 507823
Encompass_Preferred
PIK-75 HCl is a p110? inhibitor with IC50 of 5.8 nM (200-fold more potently than p110?), isoform-specific mutants at Ser773, and also potently inhibits DNA-PK with IC50 of 2 nM in cell-free assays. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-782-3 Supplier Selleck Chemical LLC Supplier No. S120510MG
PIK-75 HCl is a p110? inhibitor with IC50 of 5.8 nM (200-fold more potently than p110?), isoform-specific mutants at Ser773, and also potently inhibits DNA-PK with IC50 of 2 nM in cell-free assays. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.