Learn More
Selleck Chemical LLC PIK-90 5mg 677338-12-4
SDP
Supplier:  Selleck Chemical LLC S11875MG
Encompass_Preferred
PIK-90 is a PI3K?/?/? inhibitor with IC50 of 11 nM/18 nM/58 nM, respectively, less potent to PI3K?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-890-1